Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM3, Mouse, Biotin, Clone: MEM-171, Abnova™

Catalog No. 89112067 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-067 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-067 Supplier Abnova Corporation Supplier No. MAB5036
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ICAM3.

The protein encoded by this gene is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. This protein is constitutively and abundantly expressed by all leucocytes and may be the most important ligand for LFA-1 in the initiation of the immune response. It functions not only as an adhesion molecule, but also as a potent signalling molecule. [provided by RefSeq

Specifications

Antigen ICAM3
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-171
Conjugate Biotin
Description Mouse monoclonal antibody raised against native ICAM3.
Dilution Flow Cytometry (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ICAM3
Gene Alias CD50/CDW50/ICAM-R
Gene Symbols ICAM3
Host Species Mouse
Immunogen Native purified ICAM3 from human granulocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3385
Test Specificity This antibody MEM-171 recognizes an epitope in the D2 domain of CD50 (ICAM-3), a 120-130 KDa type I membrane protein (immunoglobulin supergene family) expressed on leukocytes, endothelial cells and Langerhans cells; it is negative on platelets and erythrocytes.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.