Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS, Mouse, Clone: ANC6C6-A3, Abnova™

Catalog No. 89113105 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-105 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-105 Supplier Abnova Corporation Supplier No. MAB6976
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant ICOS.

The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family. It forms homodimers and plays an important role in cell-cell signaling, immune responses, and regulation of cell proliferation. [provided by RefSeq

Specifications

Antigen ICOS
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone ANC6C6-A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant ICOS.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate)
Gene ICOS
Gene Alias AILIM/CD278/MGC39850
Gene Symbols ICOS
Host Species Mouse
Immunogen A mixture of human HPB-MLT cells and recombinant human ICOS-muIg.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29851
Test Specificity Antibody ANC6C6 blocks binding of recombinant CD275-muIg to CD278.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.