Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-IDH1/2 Mutant (R132/172), Clone: MsMAb-1,

Catalog No. MABC1103MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1103MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1103MI Supplier MilliporeSigma™ Supplier No. MABC1103
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects IDH1/2 Mutant (R132/172) clone: MsMAb-1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Isocitrate dehydrogenase (IDH) catalyzes the oxidative decarboxylation of isocitrate to produce alpha-ketoglutarate and carbon dioxide. There exist three forms of IDHs in human (IDH1-3), with IDH3 utilizing NAD+ as a cofactor and functioning within the citric acid cycle, while IDH1 (EC 1.1.1.42; UniProt P48735) and IDH2 (EC 1.1.1.42; UniProt P48735) utilizing NADP+ as a cofactor and functioning outside the context of the citric acid cycle. Mutated IDH1 and IDH2 convert alpha-ketoglutarate to oncometabolite R(-)-2-hydroxyglutarate (2-HG) in cytosol and mitochondria, respectively. Isocitrate dehydrogenase 1 ⁄ 2 mutations have been reported in gliomas, acute myeloid leukemias (AMLs), cartilaginous tumors, osteosarcoma, Giant cell tumors of bone (GCTB), Ollier disease, and Maffucci syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH1/2 Mutant (R132/172)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MsMAb-1
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O75874 (IDH1); P48735 (IDH2)
Gene Symbols IDH1; PICD; IDH2
Host Species Mouse
Immunogen KLH-conjugated peptide derived from human IDH1 sequence with R132G mutation.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) IDH1: NP_001269315; IDH2: NP_001276839
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.