Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Ido1, Clone: 8G11,

Catalog No. MABS485MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS485MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS485MI Supplier MilliporeSigma™ Supplier No. MABS485
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Ido1 clone: 8G11 in Mouse samples, and it is validated for Flow Cytometry, Western Blotting

Indoleamine 2,3-dioxygenase (IDO) is an enzyme that is responsible for converting tryptophan to kynurenines. IDO is expressed by a wide variety of tissues and can be upregulated by interferon gamma. IDO modulates levels of the amino acid tryptophan, which is vital for cell growth, but is also involved in the suppression of the immune response. IDO may be involved in the suppression of the immune response to tumours and blocking the IDO pathway may be a potential target for immunotherapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ido1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 8G11
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P14902
Gene Symbols IDO1; IDO; INDO
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse Ido1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002155
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.