Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI16, Mouse, Clone: 2C2, Abnova™

Catalog No. 89104715 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-715 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-715 Supplier Abnova Corporation Supplier No. H00003428M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant IFI16.

This gene encodes a member of the HIN-200 (hematopoietic interferon-inducible nuclear antigens with 200 amino acid repeats) family of cytokines. The encoded protein contains domains involved in DNA binding, transcriptional regulation, and protein-protein interactions. The protein localizes to the nucleoplasm and nucleoli, and interacts with p53 and retinoblastoma-1. It modulates p53 function, and inhibits cell growth in the Ras/Raf signaling pathway. [provided by RefSeq

Sequence: FVNGVFEVHKKNVRGEFTYYEIQDNTGKMEVVVHGRLTTINCEEGDKLKLTCFELAPKSGNTGELRSVIHSHIKVIKTRKNKKDILNPDSSMETSPDFFF

Specifications

Antigen IFI16
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant IFI16.
Formulation ascites with no preservative
Gene IFI16
Gene Accession No. BC017059
Gene Alias IFNGIP1/MGC9466/PYHIN2
Gene Symbols IFI16
Host Species Mouse
Immunogen IFI16 (AAH17059,630 a.a. ∽ 729 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3428
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.