Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFI27, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant IFI27.
Supplier: Abnova Corporation H00003429A01
Description
Sequence: MEASALTSSAVTSVAKVVRVASGSAVVLPLARIATVVIGGVVAVPMVLSAMGFTAAGIASSSIAAKMMSAAAIANGGGVASGSLVATLQSLGATGLSGLTKFILGSIGSAIAAVIARFYSpecifications
IFI27 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant IFI27. | |
IFI27 | |
FAM14D/ISG12/ISG12A/P27 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC015492 | |
IFI27 | |
IFI27 (AAH15492, 1 a.a. ∽ 119 a.a) full-length recombinant protein with GST tag. | |
RUO | |
3429 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction