Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-IFN-alpha 1, Clone: 2.52, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | IFN-alpha 1 |
---|---|
Clone | 2.52 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, SDS-Page, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24509402
![]() |
Novus Biologicals
NBP2450940.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
IFN-alpha 1 Monoclonal antibody specifically detects IFN-alpha 1 in Human samples. It is validated for Flow Cytometry, SDS-Page, Immunofluorescence.Specifications
IFN-alpha 1 | |
0.2 mg/mL | |
Flow Cytometry, SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
3439 | |
Purified recombinant human IFN-alpha1 | |
Primary | |
Store at 4C. |
2.52 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
IFNA1 | |
IgG1 κ | |
Protein A or G purified | |
This MAb is specific for human interferon alpha 1 and does not cross react with human interferon alpha 2. Interferons are widely used therapeutic agents because of their anti-tumor and anti-viral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon- & Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is influenced by phosphorylation of Serine 535 and 539 in the IFNAR1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title