Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-IFN-gamma, Clone: G-23, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450900.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
IFN-gamma | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 20-25kDa, identified as human interferon. This MAb is specific to human IFN- and recognizes both recombinant and native human IFN-gamma. It does not neutralize the activity of IFN-. T lymphocytes and NK cells mainly produce IFN-. It is a pleiotropic cytokine involved in the regulation of nearly all phases of immune and inflammatory responses, including the activation, growth and differentiation of T cell, B cells, macrophages, NK cells and other cell types such as endothelial cells and fibroblasts. It has weak anti-viral and anti-proliferative activity, and potentiates the antiviral and anti-tumor effects of IFN- (type I interferon). | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
G-23 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
IFNG | |
Purified recombinant human IFN-gamma | |
0.2 mg | |
Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
3458 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction