Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-IFN-gamma, Clone: SPM153, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | IFN-gamma |
---|---|
Clone | SPM153 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24509202
![]() |
Novus Biologicals
NBP2450920.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
IFN-gamma | |
0.2 mg/mL | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
3458 | |
Purified recombinant human IFN-gamma | |
Primary | |
Store at 4C. |
SPM153 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
IFNG | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 20-25kDa, identified as human interferon. This MAb is specific to human IFN- and recognizes both recombinant and native human IFN-gamma. It does not neutralize the activity of IFN-. T lymphocytes and NK cells mainly produce IFN-. It is a pleiotropic cytokine involved in the regulation of nearly all phases of immune and inflammatory responses, including the activation, growth and differentiation of T cell, B cells, macrophages, NK cells and other cell types such as endothelial cells and fibroblasts. It has weak anti-viral and anti-proliferative activity, and potentiates the antiviral and anti-tumor effects of IFN- (type I interferon). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title