Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNA1, Mouse, Clone: ST29, Abnova™

Catalog No. 89113117 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-117 Supplier Abnova Corporation Supplier No. MAB7027
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against IFNA1.

Leukocyte interferon is produced predominantly by B lymphocytes. Immune interferon (IFN-gamma; MIM 147570) is produced by mitogen- or antigen-stimulated T lymphocytes.[supplied by OMIM

Specifications

Antigen IFNA1
Applications Functional Assay
Classification Monoclonal
Clone ST29
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against IFNA1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene IFNA1
Gene Alias IFL/IFN/IFN-ALPHA/IFNA13/IFNA@/MGC138207/MGC138505/MGC138507
Gene Symbols IFNA1
Host Species Mouse
Immunogen Human IFNA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3439
Test Specificity Interferon alpha (ST29).
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.