Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA2, Mouse, Clone: 14AS, Abnova™

Catalog No. 89136522 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-522 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-522 Supplier Abnova Corporation Supplier No. MAB8446

Mouse monoclonal antibody raised against native IgA2.

Specifications

Antigen IgA2
Applications ELISA
Classification Monoclonal
Clone 14AS
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native IgA2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified IgA2 from human myeloma patient serum.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Subclass specific to human IgA2, Fc-region specific.The percentage of cross-reactivity was established in indirect ELISA using purified human IgG, IgA1, IgA2 and IgM as coating antigens at 5 mg/ml and is the following: IgG 7.9 %; IgA1 49 %; IgA2 100 %; IgM 3.0 %
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.