Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1R Mouse anti-Human, Clone: 3C8B1, Abnova™

Catalog No. 89136024 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-024 Supplier Abnova Corporation Supplier No. MAB5588
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant IGF1R.

Specifications

Antigen IGF1R
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 3C8B1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant IGF1R.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:2000) Immunohistochemistry (Frozen sections) (1:500-1:2000) The optimal working dilution should be determined by the end
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to IGF1R.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.