Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1R Mouse anti-Human, Clone: 3C8B1, Abnova™

Catalog No. 89108893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-893 Supplier Abnova Corporation Supplier No. MAB10692
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant IGF1R.

This receptor binds insulin-like growth factor with a high affinity. It has tyrosine kinase activity. The insulin-like growth factor I receptor plays a critical role in transformation events. Cleavage of the precursor generates alpha and beta subunits. It is highly overexpressed in most malignant tissues where it functions as an anti-apoptotic agent by enhancing cell survival. [provided by RefSeq

Specifications

Antigen IGF1R
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3C8B1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant IGF1R.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene IGF1R
Gene Alias CD221/IGFIR/JTK13/MGC142170/MGC142172/MGC18216
Gene Symbols IGF1R
Host Species Mouse
Immunogen Recombinant protein corresponding to human IGF1R.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.