Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP3, Mouse, Clone: 8F11, Abnova™

Catalog No. 89108895 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-895 Supplier Abnova Corporation Supplier No. MAB10694
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant IGF2BP3.

The protein encoded by this gene is primarily found in the nucleolus, where it can bind to the 5' UTR of the insulin-like growth factor II leader 3 mRNA and may repress translation of insulin-like growth factor II during late development. The encoded protein contains several KH domains, which are important in RNA binding and are known to be involved in RNA synthesis and metabolism. A pseudogene exists on chromosome 7, and there are putative pseudogenes on other chromosomes. [provided by RefSeq]

Specifications

Antigen IGF2BP3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8F11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant IGF2BP3.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene IGF2BP3
Gene Alias DKFZp686F1078/IMP-3/IMP3/KOC1/VICKZ3
Gene Symbols IGF2BP3
Host Species Mouse
Immunogen Recombinant protein corresponding to human IGF2BP3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10643
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.