Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2R, Mouse, FITC, Clone: MEM-238, Abnova™

Catalog No. 89112095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-095 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-095 Supplier Abnova Corporation Supplier No. MAB5071
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant IGF2R.

The protein encoded by this gene is a receptor for both, insulin-like growth factor 2 (IGF2) and mannose 6-phosphate (M6P). The IGF2 and M6P binding sites are located on different segments of the receptor. This receptor functions in the intracellular trafficking of lysosomal enzymes, the activation of transforming growth factor beta, and the degradation of IGF2. In mice, the homologous gene is expressed only from the maternal chromosome. [provided by RefSeq

Specifications

Antigen IGF2R
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-238
Conjugate FITC
Description Mouse monoclonal antibody raised against recombinant IGF2R.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene IGF2R
Gene Alias CD222/CIMPR/M6P-R/MPR1/MPRI
Gene Symbols IGF2R
Host Species Mouse
Immunogen Recombinant protein corresponding to IGF2R.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3482
Test Specificity This antibody recognizes an epitope between domains 2 and 5 of CD222 (IGF2 receptor), a ubiquitously expressed 250 KDa multifunctional type I transmembrane protein.
Target Species Human, Primate
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.