Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2R, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104728 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-728 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-728 Supplier Abnova Corporation Supplier No. H00003482A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant IGF2R.

The protein encoded by this gene is a receptor for both, insulin-like growth factor 2 (IGF2) and mannose 6-phosphate (M6P). The IGF2 and M6P binding sites are located on different segments of the receptor. This receptor functions in the intracellular trafficking of lysosomal enzymes, the activation of transforming growth factor beta, and the degradation of IGF2. In mice, the homologous gene is expressed only from the maternal chromosome. [provided by RefSeq

Sequence: GTNHRVQSSIAFLCGKTLGTPEFVTATECVHYFEWRTTAACKKDIFKANKEVPCYVFDEELRKHDLNPLIKLSGAYLVDDSDPDTSLFINVCRDIDTLRD

Specifications

Antigen IGF2R
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant IGF2R.
Formulation 50% glycerol
Gene IGF2R
Gene Accession No. NM_000876
Gene Alias CD222/CIMPR/M6P-R/MPR1/MPRI
Gene Symbols IGF2R
Host Species Mouse
Immunogen IGF2R (NP_000867, 121 a.a. ∽ 220 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3482
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.