Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-IgG, Clone: IVA285-1, Abnova™

Catalog No. 89136216 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-216 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-216 Supplier Abnova Corporation Supplier No. MAB6442
Add to Cart
Add to Cart

Specifications

Classification Monoclonal
Clone IVA285-1
Description Mouse monoclonal antibody raised against IgG.
Dilution Flow Cytometry Immunohistochemistry Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Immunogen Bovine IgG, light chain.
Quantity 100 μg
Primary or Secondary Primary
Test Specificity This antibody reacts with bovine IgG (light chains).
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.