Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (Fc), Mouse, FITC, Clone: EM-07, Abnova™

Catalog No. 89136783 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-783 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-783 Supplier Abnova Corporation Supplier No. MAB9827
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against IgG (Fc).

Specifications

Antigen IgG (Fc)
Applications Immunofluorescence
Classification Monoclonal
Clone EM-07
Concentration 0.1 mg/mL
Conjugate FITC
Description Mouse monoclonal antibody raised against IgG (Fc).
Dilution Immunofluorescence (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Fusion protein of human IgG Fc fragment.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.