Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG4, Mouse, Clone: 5C7, Abnova™

Catalog No. 89136532 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-532 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-532 Supplier Abnova Corporation Supplier No. MAB8456
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native IgG4.

Specifications

Antigen IgG4
Applications ELISA
Classification Monoclonal
Clone 5C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native IgG4.
Dilution ELISA (1:100000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified IgG4 from human serum.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Subclass-specific for human IgG4. No allotype restriction. Fc-region specific. 5C7 can be used for coating and as conjugate.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.