Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGHG3, Mouse, Clone: 5G12, Abnova™

Catalog No. 89113170 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-170 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-170 Supplier Abnova Corporation Supplier No. MAB7340
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against IGHG3.

Specifications

Antigen IGHG3
Applications ELISA
Classification Monoclonal
Clone 5G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against IGHG3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene IGHG3
Gene Alias DKFZp686H11213/FLJ39988/FLJ40036/FLJ40253/FLJ40587/FLJ40789/FLJ40834/IgG3/MGC45809
Gene Symbols IGHG3
Host Species Mouse
Immunogen Human IGHG3.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3502
Test Specificity Subclass-specific for human IgG3. No allotype restriction. Hinge-region specific.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.