Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGHM, Mouse, Clone: IM260, Abnova™

Catalog No. 89110274 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-274 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-274 Supplier Abnova Corporation Supplier No. MAB12131
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human IGHM.

Specifications

Antigen IGHM
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone IM260
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human IGHM.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene IGHM
Gene Alias DKFZp686I15196/DKFZp686I15212/FLJ00385/MGC104996/MGC52291/MU/VH
Gene Symbols IGHM
Host Species Mouse
Immunogen Recombinant protein corresponding to human IGHM.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3507
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.