Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGKC, Mouse, Clone: 4G7, Abnova™
Mouse monoclonal antibody raised against native IGKC.
Supplier: Abnova Corporation MAB7728
Specifications
IGKC | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
IGKC | |
IGKC | |
A mixture of light chains of human IGKC. | |
RUO | |
3514 | |
Human | |
Liquid |
ELISA, Immunohistochemistry | |
4G7 | |
Mouse monoclonal antibody raised against native IGKC. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809 | |
Mouse | |
200 μg | |
Primary | |
Specific for independent constantly expressed epitope of kappa chain. Reacts with free and heavy chain-associated kappa chains. | |
Store at 4°C. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction