Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGKC, Mouse, Clone: KLC264, Abnova™

Catalog No. 89109634 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-634 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-634 Supplier Abnova Corporation Supplier No. MAB11487
Add to Cart
Add to Cart

mouse monoclonal antibody raised against IGKC.

Specifications

Antigen IGKC
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KLC264
Conjugate Unconjugated
Description mouse monoclonal antibody raised against IGKC.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunohistochemistry (0.5-1 ug/mL for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein lysate) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene IGKC
Gene Alias HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809
Gene Symbols IGKC
Host Species Mouse
Immunogen Purified human IGKC.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3514
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.