Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGKC, Mouse, Clone: SB81a, Abnova™

Catalog No. 89111309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-309 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-309 Supplier Abnova Corporation Supplier No. MAB3281
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native IGKC.

Specifications

Antigen IGKC
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone SB81a
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native IGKC.
Dilution Immunofluorescence (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene IGKC
Gene Alias HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809
Gene Symbols IGKC
Host Species Mouse
Immunogen Native purified human IGKC.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3514
Test Specificity Reacts with human kappa light chains as demonstrated by ELISA.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.