Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGLC@, Mouse, Clone: 2G9, Abnova™

Catalog No. 89113191 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-191 Supplier Abnova Corporation Supplier No. MAB7752

Mouse monoclonal antibody raised against native IGLC@.

Specifications

Antigen IGLC@
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native IGLC@.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene IGLC@
Gene Alias IGLC
Gene Symbols IGLC@
Host Species Mouse
Immunogen A mixture of light chains of human IGLC@.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3536
Test Specificity Specific for independent constantly expressed epitope of lambda chain. Reacts with free and heavy chain-associated lambda chains.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.