Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF2, Mouse, Clone: BB27, Abnova™

Catalog No. 89112170 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-170 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-170 Supplier Abnova Corporation Supplier No. MAB5217
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against IGSF2.

Specifications

Antigen IGSF2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone BB27
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against IGSF2.
Dilution Flow Cytometry (10ul of the suggested working dilution (1:50-1:100) to label 106 cells in 100ul) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene IGSF2
Gene Alias CD101/V7
Gene Symbols IGSF2
Host Species Mouse
Immunogen Human thymic clone B12.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9398
Test Specificity This antibody recognizes human CD101. CD101 is a 140kD homodimeric cell surface glycoprotein expressed primarily by monocytes, granulocytes, dendritic cells and activated T lymphocytes. Removal of sodium azide is recommended prior to use in functional assays.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.