Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-IKBKB, Clone: 10A2,

Catalog No. MABS807MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS807MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS807MI Supplier MilliporeSigma™ Supplier No. MABS807
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects IKBKB clone: 10A2 in Human samples, and it is validated for Western Blotting

Inhibitor of kappa light polypeptide gene enhancer in B-cells (IKBKB), also known as kinase beta or IKK2/IKKB, is a member of the IKK complex which is composed of IKK-alpha, IKK-beta, IKK-gamma and IKAP. Phosphorylation of I-Kappa-B on a serine residue by the IKK complex frees NF-kB from I-Kappa-B and marks it for degradation via ubiquination. IKK-beta has been shown to activate NF-kB and phosphorylate IKB-alpha and beta. Phosphorylation of 2 sites at the activation loop of IKK-beta is essential for activation of IKK by TNF and IL1. Once activated, IKK-beta autophosphorylates which in turn decreases IKK activity and prevents prolonged activation of the inflammatory response. Additionally, IKK-beta activity can also be regulated by MEKK-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKBKB
Applications Western Blot
Classification Monoclonal
Clone 10A2
Formulation Mouse monoclonal IgG1 ascitic fluid containing 0.03% sodium azide.
Gene Accession No. O14920
Gene Symbols IKBKB; IKKB
Host Species Mouse
Immunogen Purified recombinant fragment of IKBKB expressed in E. coli.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.