Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKBKE, Mouse, Clone: 72B587, Abnova™

Catalog No. 89107852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-852 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-852 Supplier Abnova Corporation Supplier No. MAB0081

Mouse monoclonal antibody raised against partial recombinant IKBKE.

Specifications

Antigen IKBKE
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 72B587
Conjugate Unconjugated
Description inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene IKBKE
Gene Alias IKK-i, IKKE, IKKI, KIAA0151, MGC125294, MGC125295, MGC125297
Gene Symbols IKBKE
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 175-188 of human IKBKE.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9641
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.