Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 receptor, Mouse, Clone: IL1-31-113.1.1, Abnova™

Catalog No. 89136475 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-475 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-475 Supplier Abnova Corporation Supplier No. MAB8355
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial IL-1 receptor.

Specifications

Antigen IL-1 receptor
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone IL1-31-113.1.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial IL-1 receptor.
Dilution ELISA (5 ug/mL) Radioimmunoassay (20-30 ng/mL) Immunohistochemistry (10-100 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen The extracellular domain of the type I human IL-1 receptor (Genzyme Diagnostics).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies react with cell surface and soluble human IL-1 receptor; Specific epitopes are unknown, but they do partially inhibit (30-50%) binding of one another to IL-1 receptor. They do not block binding of IL-1 to receptor, and they do not cross-react with receptors for human IL-2 or IL-4.
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.