Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL15RA, Mouse, Clone: eBioJM7A4, Abnova™

Catalog No. 89109090 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-090 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-090 Supplier Abnova Corporation Supplier No. MAB10883

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against IL15RA.

The protein encoded by this gene is a cytokine receptor that specifically binds IL15 with high affinity. The receptors of IL15 and IL2 share two subunits, the IL2R beta and IL2R gamma chains. This forms the basis of many overlapping biological activities of IL15 and IL2. The IL2 receptor requires an additional IL2-specific alpha subunit for high affinity IL2 binding. This protein is structurally related to IL2R alpha, but is capable of binding IL15 with high affinity independent of other subunits, which suggests the distinct roles between IL15 and IL2. This receptor is reported to enhance cell proliferation and expression of apoptosis inhibitor BCL2L1/BCL2-XL and BCL2. Multiple alternatively spliced transcript variants of this gene have been reported. The full length sequences of only two variants encoding distinct isoforms are available. [provided by RefSeq

Specifications

Antigen IL15RA
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone eBioJM7A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against IL15RA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene IL15RA
Gene Alias MGC104179
Gene Symbols IL15RA
Host Species Mouse
Immunogen Human IL15RA.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3601
Test Specificity The monoclonal antibody reacts with IL15RA.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.