Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL16, Mouse, Clone: 398, Abnova™

Catalog No. 89112804 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-804 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-804 Supplier Abnova Corporation Supplier No. MAB6293
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant IL16.

The protein encoded by this gene is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of this gene undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen IL16
Applications ELISA
Classification Monoclonal
Clone 398
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant IL16.
Dilution Sandwich ELISA (Capture Coating Dose: 0.3 ug/well.) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene IL16
Gene Alias FLJ16806/FLJ42735/FLJ44234/HsT19289/IL-16/LCF/prIL-16
Gene Symbols IL16
Host Species Mouse
Immunogen Recombinant protein corresponding to human IL16.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3603
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.