Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18, Mouse, Clone: 2, Abnova™

Catalog No. 89113893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-893 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-893 Supplier Abnova Corporation Supplier No. MAB9935
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant IL18.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein (IL18BP) can specifically interact with this cytokine, and thus negatively regulate its biological activity. [provided by RefSeq

Specifications

Antigen IL18
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant IL18.
Dilution Western blot (1:1000) Immunohistochemistry (1:10) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene IL18
Gene Accession No. NM_001562.2
Gene Alias IGIF/IL-18/IL-1g/IL1F4/MGC12320
Gene Symbols IL18
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human IL18.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.