Learn More
IL1A, Mouse, Clone: 5G3, Abnova™
Mouse monoclonal antibody raised against full-length recombinant IL1A.
Supplier: Abnova Corporation MAB9932
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. This cytokine is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury, and thus induces apoptosis. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. It has been suggested that the polymorphism of these genes is associated with rheumatoid arthritis and Alzheimer's disease. [provided by RefSeq]
Specifications
IL1A | |
Monoclonal | |
Unconjugated | |
Western blot (1:1000) Immunohistochemistry (1:10) The optimal working dilution should be determined by the end user. | |
IL1A | |
IL-1A/IL1/IL1-ALPHA/IL1F1 | |
Mouse | |
Affinity purification | |
RUO | |
3552 | |
Human | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
5G3 | |
Mouse monoclonal antibody raised against full length recombinant IL1A. | |
In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
NM_000575 | |
IL1A | |
Recombinant protein corresponding to full length human IL1A. | |
100 μg | |
Primary | |
It can expression in HeLa whole cell lysate. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.