Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2, Mouse, Clone: 10C4, Abnova™

Catalog No. 89113349 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-349 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-349 Supplier Abnova Corporation Supplier No. MAB8098
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant IL2.

The protein encoded by this gene is a secreted cytokine that is important for the proliferation of T and B lymphocytes. The receptor of this cytokine is a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL4) and interleukin 7 (IL7). The expression of this gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests an essential role of this gene in the immune response to antigenic stimuli. [provided by RefSeq

Specifications

Antigen IL2
Applications ELISA
Classification Monoclonal
Clone 10C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant IL2.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Gene IL2
Gene Alias IL-2/TCGF/lymphokine
Gene Symbols IL2
Host Species Mouse
Immunogen Recombinant protein corresponding to human IL2.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3558
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.