Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2, Mouse, Clone: 12B11AE7, Abnova™

Catalog No. 89110699 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-699 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-699 Supplier Abnova Corporation Supplier No. MAB1901

Mouse monoclonal antibody raised against recombinant Il2.

Specifications

Antigen IL2
Applications ELISA
Classification Monoclonal
Clone 12B11AE7
Conjugate Unconjugated
Description interleukin 2
Formulation In PBS (0.08% sodium azide)
Gene Il2
Gene Symbols Il2
Host Species Mouse
Immunogen Recombinant protein corresponding to rat IL2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116562
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.