Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL21, Mouse, Clone: SB134b, Abnova™

Catalog No. 89113492 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-492 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-492 Supplier Abnova Corporation Supplier No. MAB8556
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant IL21.

Specifications

Antigen IL21
Applications ELISA
Classification Monoclonal
Clone SB134b
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant IL21.
Dilution ELISA (0.1-5 ug/mL) Immunostaining (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In BBS, pH 8.2
Gene IL21
Gene Alias IL-21/Za11
Gene Symbols IL21
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human IL21.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59067
Test Specificity This antibody is specific to IL21.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.