Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2RA, Mouse, Clone: TP1/6, Abnova™

Catalog No. 89112024 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-024 Supplier Abnova Corporation Supplier No. MAB4974
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native IL2RA.

The interleukin 2 (IL2) receptor alpha (IL2RA) and beta (IL2RB) chains, together with the common gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric alpha chains (IL2RA) result in low-affinity receptor, while homodimeric beta (IL2RB) chains produce a medium-affinity receptor. Normally an integral-membrane protein, soluble IL2RA has been isolated and determined to result from extracellular proteolyisis. Alternately-spliced IL2RA mRNAs have been isolated, but the significance of each is presently unknown. [provided by RefSeq

Specifications

Antigen IL2RA
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone TP1/6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native IL2RA.
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 1% BSA, pH 7.2 (0.09% sodium azide)
Gene IL2RA
Gene Alias CD25/IDDM10/IL2R/TCGFR
Gene Symbols IL2RA
Host Species Mouse
Immunogen PHA activated human T cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.