Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL31, Mouse, Clone: SB118b, Abnova™

Catalog No. 89113388 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-388 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-388 Supplier Abnova Corporation Supplier No. MAB8221
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant IL31.

IL31, which is made principally by activated Th2-type T cells, interacts with a heterodimeric receptor consisting of IL31RA (MIM 609510) and OSMR (MIM 601743) that is constitutively expressed on epithelial cells and keratinocytes. IL31 may be involved in the promotion of allergic skin disorders and in regulating other allergic diseases, such as asthma (Dillon et al., 2004 [PubMed 15184896]).[supplied by OMIM

Specifications

Antigen IL31
Applications ELISA
Classification Monoclonal
Clone SB118b
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant IL31.
Dilution ELISA (0.1-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.0
Gene IL31
Gene Alias IL-31
Gene Symbols IL31
Host Species Mouse
Immunogen Recombinant protein corresponding to human TNFSF10.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 386653
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.