Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL31RA, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-611 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-611 Supplier Abnova Corporation Supplier No. H00133396B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human IL31RA protein.

IL31RA is related to gp130 (IL6ST; MIM 600694), the common receptor subunit for IL6 (MIM 147620)-type cytokines. Oncostatin M receptor (OSMR; MIM 601743) and IL31RA form the heterodimeric receptor through which IL31 (MIM 609509) signals. Expression of IL31RA and OSMR mRNA is induced in activated monocytes, and both mRNAs are constitutively expressed in epithelial cells (Dillon et al., 2004 [PubMed 15184896]).[supplied by OMIM

Sequence: MMWTWALWMLPSLCKFSLAALPAKPENISCVYYYRKNLTCTWSPGKETSYTQYTVKRTYAFGEKHDNCTTNSSTSENRASCSFFLPRITIPDNYTIEVEAENGDGVIKSHMTYWRLENIAKTEPPKIFRVKPVLGIKRMIQIEWIKPELAPVSSDLKYTLRFRTVNSTSWMEVNFAKNRKDKNQTYNLTGLQPFTEYVIALRCAVKESKFWSDWSQEKMGMTEEEAPCGLELWRVLKPAEADGRRPVRLLWKKARGAPVLEKTLGYNIWYYPESNTNLTETMNTTNQQLELHLGGESFWVSMISYNSLGKSPVATLRIPAIQEKSFQCIEVMQACVAEDQLVVKWQSSALDVNTWMIEWFPDVDSEPTTLSWESVSQATNWTIQQDKLKPFWCYNISVYPMLHDKVGEPYSIQAYAKEGVPSEGPETKVENIGVKTVTITWKEIPKSERKGIICNYTIFYQAEGGKGFSKTVNSSILQYGLESLKRKTSYIVQVMASTSAGGTNGTSINFKTLSFSVFEIILITSLIGGGLLILIILTVAYGLKKPNKLTHLCWPTVPNPAESSIATWHGDDFKDKLNLKESDDSVNTEDRILKPCSTPSDKLVIDKLVVNFGNVLQEIFTDEARTGQENNLGGEKNGYVTCPFRPDCPLGKSFEELPVSPEIPPRKSQYLRSRMPEGTRPEAKEQLLFSGQSLVPDHLCEEGAPNPYLKNSVTAREFLVSEKLPEHTKGEV

Specifications

Antigen IL31RA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human IL31RA protein.
Formulation PBS with no preservative; pH 7.4
Gene IL31RA
Gene Accession No. NM_139017.2
Gene Alias CRL/CRL3/GLM-R/GLMR/GPL/IL-31RA/MGC125346/PRO21384
Gene Symbols IL31RA
Host Species Mouse
Immunogen IL31RA (NP_620586.2,1 a.a. ∽ 732 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133396
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.