Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL33, Mouse, Clone: 4E9, Abnova™

Catalog No. 89109463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-463 Supplier Abnova Corporation Supplier No. MAB1130
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human IL33.

Specifications

Antigen IL33
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human IL33.
Dilution ELISA Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene IL33
Gene Accession No. NM_033439
Gene Alias C9orf26/DKFZp586H0523/DVS27/NF-HEV/NFEHEV/RP11-575C20.2
Gene Symbols IL33
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 112-270 of human IL33.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90865
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.