Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL33, Mouse, Clone: SB127c, Abnova™

Catalog No. 89113389 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-389 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-389 Supplier Abnova Corporation Supplier No. MAB8222
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant IL33.

Specifications

Antigen IL33
Applications ELISA
Classification Monoclonal
Clone SB127c
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant IL33.
Dilution ELISA (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene IL33
Gene Alias C9orf26/DKFZp586H0523/DVS27/NF-HEV/NFEHEV/RP11-575C20.2
Gene Symbols IL33
Host Species Mouse
Immunogen Recombinant protein corresponding to human FASLG.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90865
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.