Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMMP2L, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-423 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-423 Supplier Abnova Corporation Supplier No. H00083943A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant IMMP2L.

The mitochondrial inner membrane peptidase (IMP) complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 (IMMP1L; MIM 612323) and IMP2 are the catalytic subunits of the IMP complex (Burri et al., 2005 [PubMed 15814844]).[supplied by OMIM

Sequence: DRVACVARVEGASMQPSLNPGGSQSSDVVLLNHWKVRNFEVHRGDIVSLVSPKNPEQKIIKRVIALEGDIVR

Specifications

Antigen IMMP2L
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant IMMP2L.
Formulation 50% glycerol
Gene IMMP2L
Gene Accession No. NM_032549
Gene Alias IMP2/IMP2-LIKE
Gene Symbols IMMP2L
Host Species Mouse
Immunogen IMMP2L (NP_115938, 31 a.a. ∽ 102 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83943
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.