Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ina, Mouse, Clone: 1D2, Abnova™

Catalog No. 89110926 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-926 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-926 Supplier Abnova Corporation Supplier No. MAB2316
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Ina.

Specifications

Antigen Ina
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 1D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Ina.
Dilution The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant
Gene Ina
Gene Alias Inexa/Intlaa/Nf66
Gene Symbols Ina
Host Species Mouse
Immunogen Recombinant protein corresponding to rat Ina.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24503
Test Specificity This antibody specifically recognizes the C-terminal non-helical extension of a-internexin.
Target Species Human, Mammalia
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.