Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INADL, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106240 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-240 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-240 Supplier Abnova Corporation Supplier No. H00010207A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant INADL.

This gene encodes a protein with multiple PDZ domains. PDZ domains mediate protein-protein interactions, and proteins with multiple PDZ domains often organize multimeric complexes at the plasma membrane. This protein localizes to tight junctions and to the apical membrane of epithelial cells. A similar protein in Drosophila is a scaffolding protein which tethers several members of a multimeric signaling complex in photoreceptors. [provided by RefSeq

Sequence: TLDTQIADDAELQKYSKLLPIHTLRLGVEVDSFDGHHYISSIVSGGPVDTLGLLQPEDELLEVNGMQLYGKSRREAVSFLKEVPPPFTLVCCRRLFDDEASVDEPRR

Specifications

Antigen INADL
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant INADL.
Formulation 50% glycerol
Gene INADL
Gene Accession No. NM_170605
Gene Alias Cipp/FLJ26982/InaD-like/PATJ
Gene Symbols INADL
Host Species Mouse
Immunogen INADL (NP_733750, 545 a.a. ∽ 651 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10207
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.