Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A haemagglutinin H7 (H7N1), Mouse, Clone: 10C6, Abnova™

Catalog No. 89136586 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-586 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-586 Supplier Abnova Corporation Supplier No. MAB8534
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Influenza A haemagglutinin H7 (H7N1).

Specifications

Antigen Influenza A haemagglutinin H7 (H7N1)
Applications Agglutination
Classification Monoclonal
Clone 10C6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Influenza A haemagglutinin H7 (H7N1).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Influenza virus A/FPV (H7N1).
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.