Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Virus Haemagglutinin H5, Mouse, Clone: 1E7D8, Abnova™

Catalog No. 89135219 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-219 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-219 Supplier Abnova Corporation Supplier No. MAB2765
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Influenza A Virus Haemagglutinin H5.

Specifications

Antigen Influenza A Virus Haemagglutinin H5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E7D8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Influenza A Virus Haemagglutinin H5.
Dilution ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to internal region 13 amino acids of Avian Influenza Hemagglutinin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.