Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Virus Haemagglutinin H5, Mouse, Clone: B200M, Abnova™

Catalog No. 89135820 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-820 Supplier Abnova Corporation Supplier No. MAB4826
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Influenza A Virus Haemagglutinin H5.

Specifications

Antigen Influenza A Virus Haemagglutinin H5
Applications ELISA
Classification Monoclonal
Clone B200M
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Influenza A Virus Haemagglutinin H5.
Dilution ELISA (detect 10ng of free peptide at 1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to 14 amino acids near internal region of IInfluenza A Virus Haemagglutinin H5 (Avian) Hemagglutinin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Hemagglutinin protein from the H5N1 strain of avian influenza (Middle Region).
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.