Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Virus Hemagglutinin H3, Mouse, Clone: InA246, Abnova™

Catalog No. 89134914 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-914 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-914 Supplier Abnova Corporation Supplier No. MAB1271
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Influenza A Virus Hemagglutinin H3.

Specifications

Antigen Influenza A Virus Hemagglutinin H3
Applications ELISA, Western Blot
Classification Monoclonal
Clone InA246
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Influenza A Virus Hemagglutinin H3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.1% gelatin, 0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to influenza A virus H3 (H3N2).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store the kit at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.