Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A virus nucleoprotein, Mouse, Clone: BDI385, Abnova™

Catalog No. 89135818 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-818 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-818 Supplier Abnova Corporation Supplier No. MAB4824
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against influenza A virus nucleoprotein.

Specifications

Antigen Influenza A virus nucleoprotein
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone BDI385
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against influenza A virus nucleoprotein.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Influenza A virus infected cell lysate.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Influenza A. specific to Nucleoprotein.
Target Species Virus
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.