Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza B virus, Mouse, Clone: FluBb, Abnova™

Catalog No. 89136596 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-596 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-596 Supplier Abnova Corporation Supplier No. MAB8553

Mouse monoclonal antibody raised against full-length recombinant influenza B virus.

Specifications

Antigen Influenza B virus
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone FluBb
Conjugate Unconjugated
Formulation In BBS, pH 8.2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length influenza virus type B matrix protein (M1).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Influenza virus type B matrix protein (M1).
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.